Learn More
Enzo Life Sciences LEADING LIGHT Wnt Reporter Cell Line (1 ml)
SDP
Catalog No. NC0648868
Encompass_Preferred
Wnt reporter cell line for discovering Wnt pathway modulators. The LEADING LIGHT. .The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway..Note: the complete LEADING LIGHT Wnt Reporter Assay Starter Kit (Prod. No. ENZ-61001) is available. Alternative name: Wnt reporter cell line. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C
Catalog No. NC0648868 Supplier Enzo Life Sciences Supplier No. ENZ610020001
Wnt reporter cell line for discovering Wnt pathway modulators. The LEADING LIGHT. .The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway..Note: the complete LEADING LIGHT Wnt Reporter Assay Starter Kit (Prod. No. ENZ-61001) is available. Alternative name: Wnt reporter cell line. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C