Learn More
Enzo Life Sciences LEADING LIGHT Wnt Cell Line Assay Reagents (1 Pack)
SDP
Catalog No. NC0912529
Encompass_Preferred
For use with the LEADING LIGHT. .The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway.The LEADING LIGHT.Note: the complete LEADING LIGHT Wnt Reporter Assay Starter Kit (Prod. No. ENZ-61001) is available. Alternative name: Wnt reporter reagents. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C
Catalog No. NC0912529 Supplier Enzo Life Sciences Supplier No. ENZ600040001
For use with the LEADING LIGHT. .The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway.The LEADING LIGHT.Note: the complete LEADING LIGHT Wnt Reporter Assay Starter Kit (Prod. No. ENZ-61001) is available. Alternative name: Wnt reporter reagents. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C