Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Leaf Pack Experiments Stream Ecology Testing Kit
SDPSCI_ED

Catalog No. s66994 Shop All LaMotte Company Products
Change view
Click to view available options
Product Type:
Leaf Pack Stream Ecology Kit
Sorting Sheet
Includes:
6 waterproof illustrated sorting sheets
Six mesh bags with labels, one waterproof marker, one Stainless Steel strainer, sorting trays 14, Plastic petri dishes 60, six waterproof illustrated sorting sheets, two thermometers, one hand held scale, two rulers, brushes 12 spoons 12, one set of 18 full color flashcards, six hand lenses, one MacroLens, one Nylon twine, six zipper top bags, one tree finder booklet, one instructions manual with record sheets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
S66994 Leaf Pack Stream Ecology Kit Six mesh bags with labels, one waterproof marker, one Stainless Steel strainer, sorting trays 14, Plastic petri dishes 60, six waterproof illustrated sorting sheets, two thermometers, one hand held scale, two rulers, brushes 12 spoons 12, one set of 18 full color flashcards, six hand lenses, one MacroLens, one Nylon twine, six zipper top bags, one tree finder booklet, one instructions manual with record sheets
S98240 Sorting Sheet 6 waterproof illustrated sorting sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog No. S66994 Supplier LaMotte™ Supplier No. 5882
Add to Cart
Add to Cart
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Includes

Six mesh bags with labels, one waterproof marker, one stainless-steel strainer, 14 sorting trays, 60 plastic Petri dishes, six waterproof illustrated sorting sheets, two thermometers, one hand-held scale, two rulers, 12 brushes, 12 spoons, one set of 18 full-color flashcards, six hand lenses, one MacroLens, one nylon twine, six zipper-top bags, one tree finder booklet, one instructions manual with record sheets

Ideal teacher demonstration is reusable, flexible and adaptable to varying time constraints and numbers of students.

Students will learn to design, implement and analyze a scientific investigation while discovering the value of aquatic macroinvertebrates as living indicators of water quality.

  • Has all the apparatus and guides necessary for collecting, sorting and identifying macroinvertebrates
  • Comprehensive instructor manual has a valuable reference guide that includes background material on stream ecology, a glossary, diagrammed instructions, experiment ideas, field lab sheets, and lab data sheets

Specifications

Grade 3+
For Use With (Application) For teacher demonstration is reusable, flexible and adaptable to varying time constraints and numbers of students
Includes Six mesh bags with labels, one waterproof marker, one Stainless Steel strainer, sorting trays 14, Plastic petri dishes 60, six waterproof illustrated sorting sheets, two thermometers, one hand held scale, two rulers, brushes 12 spoons 12, one set of 18 full color flashcards, six hand lenses, one MacroLens, one Nylon twine, six zipper top bags, one tree finder booklet, one instructions manual with record sheets
Class Size 1 Class
Product Type Leaf Pack Stream Ecology Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.