Learn More
Leica Biosystems Leaf spring - replacement for Leica microtome (compatible with RM2145)
Catalog No. NC3361632
Encompass_Preferred
Leaf Spring (MPN 14050229557) is a replacement service part from Leica Biosystems used in microtome cutting assemblies to restore correct tension and maintain sectioning performance during repairs and routine maintenance.
  • Restores correct tension in the cutting assembly.
  • Helps maintain consistent sectioning performance.
  • Intended for service and repair use.
  • Designed for straightforward replacement following service instructions.
Catalog No. NC3361632 Supplier Leica Biosystems Supplier No. 14050229557
Leaf Spring (MPN 14050229557) is a replacement service part from Leica Biosystems used in microtome cutting assemblies to restore correct tension and maintain sectioning performance during repairs and routine maintenance.
  • Restores correct tension in the cutting assembly.
  • Helps maintain consistent sectioning performance.
  • Intended for service and repair use.
  • Designed for straightforward replacement following service instructions.