Learn More
Andwin Scientific LEAK DECTECTING SOLUTION
SDP
Catalog No. NC9326287
Encompass_Preferred
Snoop Leak Detecting Solution Suitable for leak testing on any pressurized gas system including carrier gas lines and fittings and injector/detector fittings. The squeezable 8oz. bottle comes with a Sniffer Tube extendible up to 30.5cm (12 )., 1/EA
Catalog No. NC9326287 Supplier Andwin Scientific Supplier No. 32031200
Snoop Leak Detecting Solution Suitable for leak testing on any pressurized gas system including carrier gas lines and fittings and injector/detector fittings. The squeezable 8oz. bottle comes with a Sniffer Tube extendible up to 30.5cm (12 )., 1/EA