Learn More
Waters Corp Waters Corporation Leak Detection Kit, AutoPurification System, Multi-Sensor, Drip Tray
Catalog No. 50791682 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit detects leaks in drip trays of AutoPurification System modules. It features leak detection sensors and a module that acts as a junction box for additional sensors.
  • Detects leaks in various module drip trays.
  • Leak sensor module functions as a central junction box.
  • Connects up to seven additional leak sensors.
  • Includes a sensor for the SFO module drip tray.
  • Interconnect plate assembly mounts on SFO rear panel.
  • Plug connectors for external leak detectors.
  • Drip tray assembly collects potential leaks.
  • Event cable connects leak detector out to input.
  • Applicable to FractionLynx, AutoPurification, and SFO systems.
Catalog No. 50-791-682 Supplier Waters Corp Supplier No. 205000443
Add to Cart
Add to Cart
Request a Quote
This kit detects leaks in drip trays of AutoPurification System modules. It features leak detection sensors and a module that acts as a junction box for additional sensors.
  • Detects leaks in various module drip trays.
  • Leak sensor module functions as a central junction box.
  • Connects up to seven additional leak sensors.
  • Includes a sensor for the SFO module drip tray.
  • Interconnect plate assembly mounts on SFO rear panel.
  • Plug connectors for external leak detectors.
  • Drip tray assembly collects potential leaks.
  • Event cable connects leak detector out to input.
  • Applicable to FractionLynx, AutoPurification, and SFO systems.