Learn More
Electron Microscopy Sciences EMSClick Leak Proof Container, 20 ml
SDP
Catalog No. 501926591
Encompass_Preferred
The EMSClick container produces an audible ""clicking"" sound when the leak-proof seal has been properly engaged, assuring accurate closure and specimen integrity during transport. Consistent vial to vial performance is ensured by 100% virgin medical grade plastic (HDPE). The cap is designed to enhance seal during positive and negative pressure differentials and vial "ears" prevent cap from backing off during rough transport. The vials are 20 ml in capacity and are 3.04 cm in diameter and 4.43 cm in height.
Catalog No. 50-192-6591 Supplier Electron Microscopy Sciences Supplier No. 6423309
The EMSClick container produces an audible ""clicking"" sound when the leak-proof seal has been properly engaged, assuring accurate closure and specimen integrity during transport. Consistent vial to vial performance is ensured by 100% virgin medical grade plastic (HDPE). The cap is designed to enhance seal during positive and negative pressure differentials and vial "ears" prevent cap from backing off during rough transport. The vials are 20 ml in capacity and are 3.04 cm in diameter and 4.43 cm in height.