Learn More
Waters Corp WATERS CORPORATION Leak Sensor Assembly, For 25x5 Pumps, ACQUITY Modules
Catalog No. 50791687 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement Leak Sensor Assembly is designed for 25x5 pumps and ACQUITY modules. When properly configured, it is designed to stop solvent flow if solvent accumulation is detected in the surrounding reservoir.
  • Stops solvent flow upon detection of solvent accumulation in the surrounding reservoir
  • Compatible with 25x5 pumps and ACQUITY modules
Catalog No. 50-791-687 Supplier Waters Corp Supplier No. 205000505
This replacement Leak Sensor Assembly is designed for 25x5 pumps and ACQUITY modules. When properly configured, it is designed to stop solvent flow if solvent accumulation is detected in the surrounding reservoir.
  • Stops solvent flow upon detection of solvent accumulation in the surrounding reservoir
  • Compatible with 25x5 pumps and ACQUITY modules