Learn More
Description
Requires
Two AAA batteries
- Gets students actively engaged
- Features four different colors; blue, green, orange, purple
- Fun sounds: horn honk, boxing bell, doorbell and boing
- 3.5 in. in diameter
- Requires two AAA batteries (not included)
- Grades K and up
Specifications
Specifications
| Color | Blue, Green, Orange, Purple |
| Grade Level | Elementary |
| Grade | K |
| Includes | Four game show-like buzzers |
| Requires | Two AAA batteries |
| Size | 3.5 in. dia. |
| Diameter (English) Outer | 3.5 in. |
| Description | Answer Buzzers |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
