Learn More
Description
Familiar triangle format offers a powerful method for building number fact fluency and teaching fact families.
- Focuses on both addition/subtraction (on front side) and multiplication/division (on reverse)
- Supports development of higher-level algebraic skills such as equation building, the commutative property and problem solving
- Write & Wipe surface on both sides of the board
Order Info
Grades 1+. Triangle boards measure 10 in. H.
Specifications
Specifications
| Grade Level | Elementary |
| Grade | 1 |
| Height (English) | 10 in. |
| For Use With (Application) | Building number fact fluency and teaching fact families |
| Description | Write and Wipe Fact Family Boards |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
