Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Grain Cowhide Leather Drivers Gloves

Catalog No. 17200398 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-398 X-Large
17-200-395 Small
17-200-396 Medium
17-200-397 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-200-398 Supplier Wells Lamont™ Supplier No. Y0123XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grain Cowhide Leather Drivers Gloves are made from high quality cowhide to provide comfort and protection while on the job. A double shirred elastic wrist provides a secure fit.

  • Quality grain cowhide leather
  • Double shirred elastic wrist
  • Bound hem
  • Straight thumb
  • Gunn cut

Specifications

Cuff Material Elastic
Color Natural
Hand Specific
Latex Free Yes
Material Grain Cowhide Leather
Product Type Driver Gloves
Size X-Large
Style Gunn
For Use With (Application) Warehousing, Construction, Oil and Gas
Gender Unisex
Glove Cuff Color Blue
Glove Cuff Style Open
Hem Color Blue
Series Y0123
Thumb Style Straight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.