Learn More
MSC Fibre-Metal Welding Helmet, Leather, Lift-Front, Shade 10, 1 ea
Catalog No. 5000508281
Encompass
This lift-front welding helmet is constructed from durable red leather, providing reliable arc glare protection and long-lasting performance in high-heat environments. Designed specifically for use in confined or hard-to-reach areas where standard helmets are impractical, it features a flexible leather construction that conforms comfortably to the user.
  • Compact design suitable for confined-space welding applications
  • Lift-front glassholder allows for rapid inspection and grinding without headgear removal
  • Screened ventilation ports facilitate improved airflow
  • Adjustable overhead headband enables optimal viewing and a comfortable fit
  • Includes a 2 in. x 4 1/4 in. size 10 shade lens
  • ANSI and CSA compliant
Catalog No. 50-005-08281 Supplier MSC Supplier No. 45845237
This lift-front welding helmet is constructed from durable red leather, providing reliable arc glare protection and long-lasting performance in high-heat environments. Designed specifically for use in confined or hard-to-reach areas where standard helmets are impractical, it features a flexible leather construction that conforms comfortably to the user.
  • Compact design suitable for confined-space welding applications
  • Lift-front glassholder allows for rapid inspection and grinding without headgear removal
  • Screened ventilation ports facilitate improved airflow
  • Adjustable overhead headband enables optimal viewing and a comfortable fit
  • Includes a 2 in. x 4 1/4 in. size 10 shade lens
  • ANSI and CSA compliant