Learn More
Medchemexpress LLC Leb-03-146 | 2858812-91-4 | 98.1% | C46H57N11O8 | 1 MG
SDP
Catalog No. 5000354617
Encompass_Preferred
LEB-03-146 is a deubiquitinase-targeting chimera (DUBTAC) that links AZD1775 (Adavosertib) to the OTUB1 recruiter EN523 via a PEG2 linker. This compound is intended for research use only.
  • Demonstrates significant WEE1 stabilization in HEP3B hepatoma cancer cells.
  • Functions as a deubiquitinase-targeting chimera (DUBTAC).
  • Links AZD1775 (Adavosertib) to the OTUB1 recruiter EN523.
Catalog No. 50-003-54617 Supplier Medchemexpress LLC Supplier No. HY1449991MG
LEB-03-146 is a deubiquitinase-targeting chimera (DUBTAC) that links AZD1775 (Adavosertib) to the OTUB1 recruiter EN523 via a PEG2 linker. This compound is intended for research use only.
  • Demonstrates significant WEE1 stabilization in HEP3B hepatoma cancer cells.
  • Functions as a deubiquitinase-targeting chimera (DUBTAC).
  • Links AZD1775 (Adavosertib) to the OTUB1 recruiter EN523.