Learn More
Research Products International Corp Lecithin, Refined From Soybeans, 500 Grams

Supplier: Research Products International Corp L46100500.0
- CAS Number: 8002-43-5
- Soluble in: Chloroform
- Store at: -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.