Learn More
NEOGEN Lecithin, 500 g
SDP
Supplier:  NEOGEN 700004725
Encompass_Preferred
Lecithin is used when preparing microbiology culture media in a laboratory setting, and is not intended for use in the diagnosis of disease or other conditions in humans.A deoiled, powdered soybean lecithin for use in microbiological culture media. A mixture of polar (phospho- and glyco-) lipids and a small amount of carbohydrates.For more information visit https://www.neogen.com/solutions/Microbiology/lecithin
Catalog No. 50-201-5864