Learn More
Alkali Scientific Lecithin, Refined From Soybeans, 100 G
SDP
Supplier:  Alkali Scientific LEC001CP
Encompass_Preferred
Lecithin, refined from soybeans, is a versatile phospholipid commonly used as an emulsifier in food, pharmaceutical, and cosmetic formulations. This 100g quantity of lecithin is essential in formulations requiring stable emulsions, where lecithin acts as a surfactant to help oil and water mix. It is commonly used in the preparation of liposomes, which are utilized in drug delivery systems, as well as in cosmetic creams and lotions to improve texture and stability. Lecithin is also used in cell culture media for the preparation of cell membranes and as a nutrient source in various laboratory applications. This product provides a reliable source of lecithin for diverse industrial and laboratory uses.
Catalog No. 50-006-59769