Learn More
Alkali Scientific Lecithin, Refined From Soybeans, 25 G
SDP
Catalog No. 5000659761
Encompass_Preferred
Lecithin, refined from soybeans, is a naturally occurring phospholipid used widely as an emulsifier in laboratory and industrial applications. This 25g bottle is ideal for small-scale formulations, including use in cell culture media, where lecithin plays a critical role in membrane structure and function. Lecithin is also used in the preparation of liposomes, which are valuable in drug delivery and gene therapy applications. The product helps stabilize emulsions in food products, pharmaceuticals, and cosmetics. Its ability to reduce surface tension and enhance solubility makes it an essential ingredient in many formulations, offering a natural solution to create stable mixtures of oil and water-based substances.
Catalog No. 50-006-59761 Supplier Alkali Scientific Supplier No. LEC002CP
Lecithin, refined from soybeans, is a naturally occurring phospholipid used widely as an emulsifier in laboratory and industrial applications. This 25g bottle is ideal for small-scale formulations, including use in cell culture media, where lecithin plays a critical role in membrane structure and function. Lecithin is also used in the preparation of liposomes, which are valuable in drug delivery and gene therapy applications. The product helps stabilize emulsions in food products, pharmaceuticals, and cosmetics. Its ability to reduce surface tension and enhance solubility makes it an essential ingredient in many formulations, offering a natural solution to create stable mixtures of oil and water-based substances.