Learn More
Alkali Scientific Lecithin, Refined From Soybeans, 5 Kilograms
SDP
Catalog No. 5000659820
Encompass_Preferred
Lecithin, Refined from Soybeans, 5KG, is a phospholipid mixture widely used in research for its emulsifying, stabilizing, and dispersing properties. It is derived from soybeans and is rich in phosphatidylcholine, which plays a vital role in cellular membrane structure and function. This 5kg bulk package is ideal for large-scale applications, including laboratory research, pharmaceutical formulations, and food technology. Lecithin is used in the preparation of liposomal formulations, drug delivery systems, and in cosmetic products. It also has applications in food processing as an emulsifier, helping to improve the texture and consistency of products such as chocolate, margarine, and salad dressings.
Catalog No. 50-006-59820 Supplier Alkali Scientific Supplier No. LEC004CP
Lecithin, Refined from Soybeans, 5KG, is a phospholipid mixture widely used in research for its emulsifying, stabilizing, and dispersing properties. It is derived from soybeans and is rich in phosphatidylcholine, which plays a vital role in cellular membrane structure and function. This 5kg bulk package is ideal for large-scale applications, including laboratory research, pharmaceutical formulations, and food technology. Lecithin is used in the preparation of liposomal formulations, drug delivery systems, and in cosmetic products. It also has applications in food processing as an emulsifier, helping to improve the texture and consistency of products such as chocolate, margarine, and salad dressings.