Learn More
Alkali Scientific Lecithin, Refined From Soybeans, 500 G
SDP
Catalog No. 5000659842
Encompass_Preferred
Lecithin, refined from soybeans, is a high-quality phospholipid used as an emulsifier in many industrial applications, including pharmaceuticals, food processing, and cosmetic formulations. This 500g quantity is ideal for large-scale preparations, particularly in the production of emulsions, liposomes, and cell culture media. In drug formulation, lecithin helps enhance the bioavailability of poorly soluble drugs by facilitating the formation of liposomes, which are used for targeted drug delivery. The product is also utilized in the creation of stable emulsions in cosmetics and personal care products. Lecithin’s natural properties and versatile uses make it indispensable for many commercial and research-based applications.
Catalog No. 50-006-59842 Supplier Alkali Scientific Supplier No. LEC003CP
Lecithin, refined from soybeans, is a high-quality phospholipid used as an emulsifier in many industrial applications, including pharmaceuticals, food processing, and cosmetic formulations. This 500g quantity is ideal for large-scale preparations, particularly in the production of emulsions, liposomes, and cell culture media. In drug formulation, lecithin helps enhance the bioavailability of poorly soluble drugs by facilitating the formation of liposomes, which are used for targeted drug delivery. The product is also utilized in the creation of stable emulsions in cosmetics and personal care products. Lecithin’s natural properties and versatile uses make it indispensable for many commercial and research-based applications.