Learn More
BUCHI Leck-Adapter
SDP
Catalog No. 05405074 Shop All BUCHI Products
Change view
Click to view available options
Material:
Borosilicate Glass (3.3)
Catalog No. Material
05-405-074 Borosilicate Glass (3.3)
1 options
Catalog No. 05-405-074 Supplier BUCHI Supplier No. 11063430
Add to Cart
Add to Cart
Intermediate piece with valve placed between condenser and receiving flask. Includes drain valve, hose connection (8 mm I.D.), BJ 35/20, and a connection piece with 3-way valve.
  • Allows for the removal and emptying of the receiving flask during evaporation
Specifications
Material Borosilicate Glass (3.3)
Type Leck-Adapter
For Use With (Application) 35/20 Glass Ball Joint, Condenser, Receiving Flask
For Use With (Equipment) R-300, R-210, R-215, R-100, R II
Includes Drain Valve, Hose Connection (8 mm I.D.), BJ 35/20, Connection Piece with 3-Way Valve
Taper 35/20 Glass Ball Joint