Learn More
LECO Corporation Lecosorb (20/30 Mesh), 500 Grams, for CO2 Removal
Supplier:  LECO Corporation 502174HAZ
Encompass
LECOSORB,Component: Sodium Hydroxide (caustic soda),Coated on Silicate, Light green solid,BOILinG POinT: (at 760mm Hg) (1388 Degree C),VAPOR PRESSURE (mm): 42mm Hg at 1832 Degree F (1000 C),Completely soluble Water
Catalog No. NC9278554