Learn More
Medchemexpress LLC Lecozotan hydrochloride | 433282-68-9 | 98.7% | 520.02 g/mol | C28H30ClN5O3 | 10 MG
SDP
Catalog No. 5000294626
Encompass_Preferred
Lecozotan hydrochloride is the hydrochloride salt of lecozotan, a selective 5-HT1A receptor antagonist used in research to investigate serotonergic signaling and cognitive function. The compound is provided as a characterized solid for in vitro and in vivo studies, with supporting analytical documentation available.
  • High reported purity of 98.7% suitable for research applications.
  • Off-white to light-yellow solid, easy to handle and store.
  • Molecular weight 520.02 g/mol and formula C28H30ClN5O3 for precise dosing calculations.
  • Available in small milligram quantities for early-stage studies.
  • COA, data sheet, and SDS available for quality and safety verification.
Catalog No. 50-002-94626 Supplier Medchemexpress LLC Supplier No. HY19742A10MG
Lecozotan hydrochloride is the hydrochloride salt of lecozotan, a selective 5-HT1A receptor antagonist used in research to investigate serotonergic signaling and cognitive function. The compound is provided as a characterized solid for in vitro and in vivo studies, with supporting analytical documentation available.
  • High reported purity of 98.7% suitable for research applications.
  • Off-white to light-yellow solid, easy to handle and store.
  • Molecular weight 520.02 g/mol and formula C28H30ClN5O3 for precise dosing calculations.
  • Available in small milligram quantities for early-stage studies.
  • COA, data sheet, and SDS available for quality and safety verification.