Learn More
Raybiotech Inc Lectin Array 70.
Supplier:  Raybiotech Inc GALECTIN7056
Encompass_Preferred
Detects glycan profiles using 70 lectins. Suitable for all liquid sample types. Proteins detected: AAA, AAL, ACG, ACL, ASA, BanLec, BC2L-A, BC2LCN, BPA, Calsepa, CGL2, CNL, Con A, DBA, Discoidin I, Discoidin II, DSA, ECA, EEL, F17AG, Gal1, Gal1-S, Gal2, Gal3, Gal3C-S, Gal7-S, Gal9, GNA, GRFT, GS-I, GS-II, HIHA, Jacalin, LBA, LCA, LEA, Lentil, Lotus, LSL-N, MAA, Malectin, MOA, MPL, NPA, Orysata, PA-IIL, PA-IL, PALa, PHA-E, PHA-L, PHA-P, PNA, PPL, PSA, PSL1a, PTL, RS-Fuc, SAMB, SBA, SJA, SNA-I, SNA-II, STL, UDA, UEA-I, UEA-II, VFA, VVA, WFA, WGA
Catalog No. 50-195-0326