Learn More
Raybiotech Inc Lectin Array 95.
Catalog No. 501950328
Encompass_Preferred
Detects glycan profiles using 95 lectins. Suitable for all liquid sample types. Proteins detected: AAA, AAL, ABA, ABL, ACG, ACL, AMA, ASA, BanLec, BC2L-A, BC2LCN, BPA, CA, CAA, Calsepa, CGL2, CNL, Con A, CPA, CSA, DBA, Discoidin I, Discoidin II, DSA, ECA, EEL, F17AG, Gal1, Gal1-S, Gal2, Gal3, Gal3C-S, Gal7-S, Gal9, GHA, GNA, GRFT, GS-I, GS-II, HAA, HHA, HMA, IRA, Jacalin, LAL, LBA, LCA, LEA, Lentil, Lotus, LPA, LSL-N, MAA, Malectin, MNA-G, MNA-M, MOA, MPL, NPA, Orysata, PA-IIL, PA-IL, PALa, PHA-E, PHA-L, PHA-P, PNA, PPL, PSA, PSL1a, PTL-1, PTL-2, PWA, RCA-120, RCA-60, RPA, RS-Fuc, SAMB, SBA, SHA, SJA, SNA-I, SNA-II, SSA, STL, TL, UDA, UEA-I, UEA-II, VFA, VRA, VVA, VVA-M, WFA, WGA
Catalog No. 50-195-0328 Supplier Raybiotech Inc Supplier No. GALECTIN9524
Detects glycan profiles using 95 lectins. Suitable for all liquid sample types. Proteins detected: AAA, AAL, ABA, ABL, ACG, ACL, AMA, ASA, BanLec, BC2L-A, BC2LCN, BPA, CA, CAA, Calsepa, CGL2, CNL, Con A, CPA, CSA, DBA, Discoidin I, Discoidin II, DSA, ECA, EEL, F17AG, Gal1, Gal1-S, Gal2, Gal3, Gal3C-S, Gal7-S, Gal9, GHA, GNA, GRFT, GS-I, GS-II, HAA, HHA, HMA, IRA, Jacalin, LAL, LBA, LCA, LEA, Lentil, Lotus, LPA, LSL-N, MAA, Malectin, MNA-G, MNA-M, MOA, MPL, NPA, Orysata, PA-IIL, PA-IL, PALa, PHA-E, PHA-L, PHA-P, PNA, PPL, PSA, PSL1a, PTL-1, PTL-2, PWA, RCA-120, RCA-60, RPA, RS-Fuc, SAMB, SBA, SHA, SJA, SNA-I, SNA-II, SSA, STL, TL, UDA, UEA-I, UEA-II, VFA, VRA, VVA, VVA-M, WFA, WGA