Learn More
Biotium Lectin Conjugates Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF740 1/EA
Catalog No. 5000501037
Encompass_Preferred
Arachis hypogaea (peanut) PNA lectin is specific for terminal -galactose and binds preferentially to a commonly occurring structure galactosyl (-1 3) N-acetylgalactosamine Near-infrared CF 740 has Ex/Em at 745/786 nm 1 mg lyophilized solid
Catalog No. 50-005-01037 Supplier Biotium Supplier No. 29137
Arachis hypogaea (peanut) PNA lectin is specific for terminal -galactose and binds preferentially to a commonly occurring structure galactosyl (-1 3) N-acetylgalactosamine Near-infrared CF 740 has Ex/Em at 745/786 nm 1 mg lyophilized solid