Learn More
Biotium Lectin Conjugates Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF790 1/EA
Catalog No. 5000500155
Encompass_Preferred
Arachis hypogaea (peanut) PNA lectin is specific for terminal -galactose and binds preferentially to a commonly occurring structure galactosyl (-1 3) N-acetylgalactosamine Near-infrared CF790 has Ex/Em at 745/786 nm 1 mg lyophilized solid
Catalog No. 50-005-00155 Supplier Biotium Supplier No. 29143
Arachis hypogaea (peanut) PNA lectin is specific for terminal -galactose and binds preferentially to a commonly occurring structure galactosyl (-1 3) N-acetylgalactosamine Near-infrared CF790 has Ex/Em at 745/786 nm 1 mg lyophilized solid