Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Arachis hypoga25MG
Catalog No. 502904203
Encompass
Lectin from Arachis hypogaea (peanut) is a homotetrameric protein of 110 kDa.
Catalog No. 50-290-4203 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L088125MG
Lectin from Arachis hypogaea (peanut) is a homotetrameric protein of 110 kDa.