Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Arachis hypoga5MG
Catalog No. 502904543
Encompass
Lectins are carbohydrate-binding proteins omnipresent found in fungi plants and animals. The structure of lectin is diversely studied in plants and animals. The secondary structure of this protein is rich in -strands and possesses a carbohydrate binding sites on the surface.
Catalog No. 50-290-4543 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L73815MG
Lectins are carbohydrate-binding proteins omnipresent found in fungi plants and animals. The structure of lectin is diversely studied in plants and animals. The secondary structure of this protein is rich in -strands and possesses a carbohydrate binding sites on the surface.