Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Bandeiraea sim1MG
Catalog No. 502904207
Encompass
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia) has A and B subunits which recognizesGalNAc andGal-end groups respectively. Lectins are extracted from the seeds of the legume shrub Griffonia simplicifolia which are carbohydrate binding proteins.
Catalog No. 50-290-4207 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L21401MG
Add to Cart
Add to Cart
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia) has A and B subunits which recognizesGalNAc andGal-end groups respectively. Lectins are extracted from the seeds of the legume shrub Griffonia simplicifolia which are carbohydrate binding proteins.