Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Bandeiraea sim2MG
Catalog No. 502900089
Encompass
Lectins are carbohydrate-binding proteins omnipresent found in fungi plants and animals. The structure of lectin is diversely studied in plants and animals. The secondary structure of this protein is rich in -strands and possesses carbohydrate binding sites on the surface.
Catalog No. 50-290-0089 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L2895.2MG
Lectins are carbohydrate-binding proteins omnipresent found in fungi plants and animals. The structure of lectin is diversely studied in plants and animals. The secondary structure of this protein is rich in -strands and possesses carbohydrate binding sites on the surface.