Learn More
Crescent Chemical Co Inc LECTIN FROM CANAVALIA ENSIFORM
SDP
Supplier:  Crescent Chemical Co Inc SE27648.03
Encompass_Preferred
Lectin from Canavalia ensiformis lyophil. Affinity chromatography (1). Action on tumor cells (2).Sugar specificity: D-glucose, D-mannose and sterically related sugarsHemagglutination: Reference (3) Size - 1G Storage Conditions - +2 °C TO +8 °C Catalog Number - 2764
Catalog No. 50-247-799