Learn More
Crescent Chemical Co Inc LECTIN FROM CANAVALIA ENSIFORM
SDP
Catalog No. 50247797
Encompass_Preferred
Lectin from Canavalia ensiformis lyophil. Affinity chromatography (1). Action on tumor cells (2).Sugar specificity: D-glucose, D-mannose and sterically related sugarsHemagglutination: Reference (3) Size - 100MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 2
Catalog No. 50-247-797 Supplier Crescent Chemical Co Inc Supplier No. SE27648.01
Lectin from Canavalia ensiformis lyophil. Affinity chromatography (1). Action on tumor cells (2).Sugar specificity: D-glucose, D-mannose and sterically related sugarsHemagglutination: Reference (3) Size - 100MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 2