Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Galanthus niva5MG
Catalog No. 502900699
Encompass
Lectin from Galanthus nivalis (snowdrop) is a tetrameric 12 kDa mannose binding lectin. It belongs to the family Amaryllidacea. It is specific to 1 3- or 1 6- linked D-mannose of carbohydrates. Snowdrop lectins are toxic to insects.
Catalog No. 50-290-0699 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L82755MG
Lectin from Galanthus nivalis (snowdrop) is a tetrameric 12 kDa mannose binding lectin. It belongs to the family Amaryllidacea. It is specific to 1 3- or 1 6- linked D-mannose of carbohydrates. Snowdrop lectins are toxic to insects.