Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Lycopersicon e1MG
Catalog No. 502904114
Encompass
Lectins are carbohydrate binding proteins. Lectin from Lycopersicon esculentum (LEA) binds strongly to polylactosamine oligosaccharides. It is the major component of endothelial and epithelial cell surface carbohydrate.
Catalog No. 50-290-4114 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L04011MG
Lectins are carbohydrate binding proteins. Lectin from Lycopersicon esculentum (LEA) binds strongly to polylactosamine oligosaccharides. It is the major component of endothelial and epithelial cell surface carbohydrate.