Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Phaseolus vulg50MG
Catalog No. 502901146
Encompass
Phytohaemagglutinin (PHA or phytohemagglutinin) is a lectin found in plants (esp. in legumes). It is obtained from Phaseolus vulgaris (red kidney beans) and acts as a mitogen in human leucocyte culture.
Catalog No. 50-290-1146 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L875450MG
Phytohaemagglutinin (PHA or phytohemagglutinin) is a lectin found in plants (esp. in legumes). It is obtained from Phaseolus vulgaris (red kidney beans) and acts as a mitogen in human leucocyte culture.