Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Phaseolus vulg5MG
Catalog No. 502904553
Encompass
Phytohemagglutinin (PHA) derived from Phaseolus vulgaris is a tetrameric glycoprotein. It consists of two different subunits that weighs 120 kDa.
Catalog No. 50-290-4553 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L27695MG
Phytohemagglutinin (PHA) derived from Phaseolus vulgaris is a tetrameric glycoprotein. It consists of two different subunits that weighs 120 kDa.