Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Phytolacca ame10MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L877710MG
Encompass
Lectins are proteins of plant origin which have the ability to bind to certain neuronal surface glycoproteins and glycolipids. This ability of theirs helps them to be used as neuroanatomical tracers as they are taken and transported in a retrograde or anterograde fashion.
Catalog No. 50-290-1835