Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Phytolacca ame40MG
Catalog No. 502914415
Encompass
Lectins are carbohydrate-binding proteins omnipresent found in fungi plants and animals. The structure of lectin is diversely studied in plants and animals. The secondary structure of this protein is rich in -strands and possesses a carbohydrate binding sites on the surface.
Catalog No. 50-291-4415 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L937940MG
Lectins are carbohydrate-binding proteins omnipresent found in fungi plants and animals. The structure of lectin is diversely studied in plants and animals. The secondary structure of this protein is rich in -strands and possesses a carbohydrate binding sites on the surface.