Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Phytolacca ame5MG
Catalog No. 502901899
Encompass
Lectins are carbohydrate-binding proteins omnipresent found in fungi plants and animals. The structure of lectin is diversely studied in plants and animals. The secondary structure of this protein is rich in -strands and possesses a carbohydrate binding sites on the surface.
Catalog No. 50-290-1899 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L93795MG
Lectins are carbohydrate-binding proteins omnipresent found in fungi plants and animals. The structure of lectin is diversely studied in plants and animals. The secondary structure of this protein is rich in -strands and possesses a carbohydrate binding sites on the surface.