Learn More
Sigma Aldrich Fine Chemicals Biosciences Lectin from Pisum sativum2MG
Catalog No. 502900122
Encompass
Lectin from Pisum sativum (pea) is a tetrameric glycoprotein of about 48 kDa. It is isolated from peas. Pisum sativum agglutinin is specific for D-glucose and D-mannose.
Catalog No. 50-290-0122 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L07702MG
Lectin from Pisum sativum (pea) is a tetrameric glycoprotein of about 48 kDa. It is isolated from peas. Pisum sativum agglutinin is specific for D-glucose and D-mannose.