Learn More
MOLECULAR DEPOT LLC Lectin Helix p. Biotin -1 mg
Catalog No. 5000013371
Encompass_Preferred
Lectin from Helix pomatia Conjugated to Biotin is a highly purified and concentrated solution of Helix pomatia Lectin conjugated to biotin This product is provided as a lyophilized powder and has been tested for streptavidin capture For in vitro research use only
Catalog No. 50-000-13371 Supplier MOLECULAR DEPOT LLC Supplier No. B2012610
Lectin from Helix pomatia Conjugated to Biotin is a highly purified and concentrated solution of Helix pomatia Lectin conjugated to biotin This product is provided as a lyophilized powder and has been tested for streptavidin capture For in vitro research use only