Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lectin, Wheat Germ Agglutinin, MP Biomedicals™

Catalog No. MP215226605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215226605 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215226605 Supplier MP Biomedicals, Inc Supplier No. 0215226605
Add to Cart
Add to Cart

Valuable in a variety of applications, including mitogenic stimulation of lymphocytes. Lectin, Wheat Germ Agglutinin, MP Biomedicals is not blood group specific, but has an affinity for N-acetyl-β-D-glucosamine residues and oligomers. WGA contains no

Lectins are proteins or glycoproteins of non-immune origin that agglutinate cells and/or precipitate complex carbohydrates. Lectins are capable of binding glycoproteins even in presence of various detergents. The agglutination activity of these highly specific carbohydrate-binding molecules is usually inhibited by a simple monosaccharide, but for some lectins, di, tri, and even polysaccharides are required.

The lecithin in this product is purified Triticum vulgrislectin from Wheat Germ Agglutinin (WGA). It is very valuable in a wide variety of applications in vitro, including:

  • Blood grouping and erythrocyte polyagglutination studies
  • Mitogenic stimulation of lymphocytes
  • Lymphocyte subpopulation studies
  • Fractionation of cells and other particles
  • Histochemical studies of normal and pathological conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym WGA, Wheat germ agglutinin, Lectin from Triticum vulgaris (wheat)

Specifications

Quantity 5 mg
Solubility Information Dissolvable in distilled water and 0.9% NaCl within 5 minutes.
Concentration or Composition (by Analyte or Components) Working Concentration: Reconstitute with Buffer to a concentration of 1mg/mL.
Physical Form Lyophilized Powder
Recommended Storage −20°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.