Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lectin, Wheat Germ Agglutinin from Triticum vulgaris , MP Biomedical

Catalog No. MP215226625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215226625 25 mg
MP215226680 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215226625 Supplier MP Biomedicals, Inc Supplier No. 0215226625
Add to Cart
Add to Cart

Lectin, Wheat Germ Agglutinin from Triticum vulgaris are proteins or glycoproteins of non-immune origin that agglutinate cells and/or precipitate complex carbohydrates.

  • Lectins are very valuable in a wide variety of applications in vitro, including blood grouping and erythrocyte polyagglutination studies, Mitogenic stimulation of lymphocytes, Lymphocyte subpopulation studies, Fractionation of cells and other particles, Histochemical studies of normal and pathological conditions
  • Lectins are capable of binding glycoproteins even in presence of various detergents
  • The agglutination activity of these highly specific carbohydrate-binding molecules is usually inhibited by a simple monosaccharide, but for some lectins, di, tri, and even polysaccharides are required
  • It is purified Triticum vulgrislectin (WGA) from Wheat Germ
  • WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers
  • WGA contains no protein-bound carbohydrate
TRUSTED_SUSTAINABILITY

Specifications

Color White
Quantity 25 mg
Packaging Glass
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.