Learn More
Vygon USA Lectro-Cath 1.0mm ID, 50cm long , Female Luer-lock / Male Luer-lock.
SDP
Catalog No. NC1825632
Encompass_Preferred
Lectrocath 50cm micro bore male​/​female luer lock PE extension set. Polyethylene extension tube for intravenous or arterial catheters with a male Luer-lock connector at the distal end and a female Luer-lock connector at the proximal end. Lectro-Cath is supplied in a double packaging with colour-coded clip: blue for venous lines and red for arterial lines. The polyethylene material of the tube confers excellent chemical inertness limiting the interactions (adsorption, absorption...) with infused drugs.
Catalog No. NC1825632 Supplier Vygon USA Supplier No. 1155.05
Lectrocath 50cm micro bore male​/​female luer lock PE extension set. Polyethylene extension tube for intravenous or arterial catheters with a male Luer-lock connector at the distal end and a female Luer-lock connector at the proximal end. Lectro-Cath is supplied in a double packaging with colour-coded clip: blue for venous lines and red for arterial lines. The polyethylene material of the tube confers excellent chemical inertness limiting the interactions (adsorption, absorption...) with infused drugs.