Learn More
Grainger LED 1 W T3-1/4 Miniature Bayonet (BA9s)
Catalog No. 502751892
Encompass_Preferred
The Grainger Approved LED 1 Watt T3 1/4 Miniature Bulb is a 1 watt LED light bulb with a T3 1/4 miniature screw base. It produces 100 lumens of warm white light at a color temperature of 2700K. This LED bulb is rated to last 25,000 hours. It has an input voltage range of 120-277V AC. The dimensions are 1-1/4"L x 9/16"D. This LED light bulb is designed to replace a standard incandescent T3 1/4 bulb up to 25 watts. It is ideal for use in night lights, holiday lighting, appliance interior lighting and more. This miniature LED bulb is UL listed and offers an energy efficient lighting solution.
Catalog No. 50-275-1892 Supplier Grainger Supplier No. 14A148
The Grainger Approved LED 1 Watt T3 1/4 Miniature Bulb is a 1 watt LED light bulb with a T3 1/4 miniature screw base. It produces 100 lumens of warm white light at a color temperature of 2700K. This LED bulb is rated to last 25,000 hours. It has an input voltage range of 120-277V AC. The dimensions are 1-1/4"L x 9/16"D. This LED light bulb is designed to replace a standard incandescent T3 1/4 bulb up to 25 watts. It is ideal for use in night lights, holiday lighting, appliance interior lighting and more. This miniature LED bulb is UL listed and offers an energy efficient lighting solution.