Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger LED 23 W T5 4-Pin 2G11

Catalog No. 502777154
Encompass_Preferred

LED 23 W T5 4-Pin (2G11)

Catalog No. 50-277-7154 Supplier Grainger Supplier No. 184U06
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LED 23 W T5 4-Pin (2G11)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.