Learn More
Grainger LED 9 W BR30
Catalog No. 502750014
Encompass_Preferred
The TCP Elite LED 157K20 is a 15-watt LED light bulb with a medium base that produces 1,100 lumens of soft white light at a color temperature of 2700K. It has an estimated life of 25,000 hours and is dimmable. This energy-efficient bulb can replace a 60-watt incandescent bulb and is designed for indoor use in table lamps, floor lamps and more. It measures 4.25 inches tall with a diameter of 2.36 inches. This LED light bulb is UL listed, has a CRI of 80+ and contains no mercury.
Catalog No. 50-275-0014 Supplier Grainger Supplier No. 157K20
The TCP Elite LED 157K20 is a 15-watt LED light bulb with a medium base that produces 1,100 lumens of soft white light at a color temperature of 2700K. It has an estimated life of 25,000 hours and is dimmable. This energy-efficient bulb can replace a 60-watt incandescent bulb and is designed for indoor use in table lamps, floor lamps and more. It measures 4.25 inches tall with a diameter of 2.36 inches. This LED light bulb is UL listed, has a CRI of 80+ and contains no mercury.