Learn More
Microscope World SCHOTT KL 2500 LED Cold Light Source
SDP
Catalog No. NC2965463
Encompass_Preferred
This SCHOTT KL 2500 LED cold light source has electronic shutter function, LCD display indicating brightness, operation mode, or error code, and USB connectivity.Color Temperature- 5600KLuminous Flux- Up to11002 lm light flux.Voltage- 100-240V, 50-60Hz.Light Box- Intensity control 0-100% with 3 sensitivities, and filter slot with 2-position filter slider. LCD display indicates brightness / operation mode. USB connectivity for PC control. Extra-fine dimming option down to 0.1% steps.Light Box Dimensions / Weight- 114mm x 231mm x 137mm (4.5inch x 9.1inch x 5.4inch), 3.6kg (7.9 lb)Certifications- CEWarranty- 2 years
Catalog No. NC2965463 Supplier Microscope World Supplier No. KL2500
This SCHOTT KL 2500 LED cold light source has electronic shutter function, LCD display indicating brightness, operation mode, or error code, and USB connectivity.Color Temperature- 5600KLuminous Flux- Up to11002 lm light flux.Voltage- 100-240V, 50-60Hz.Light Box- Intensity control 0-100% with 3 sensitivities, and filter slot with 2-position filter slider. LCD display indicates brightness / operation mode. USB connectivity for PC control. Extra-fine dimming option down to 0.1% steps.Light Box Dimensions / Weight- 114mm x 231mm x 137mm (4.5inch x 9.1inch x 5.4inch), 3.6kg (7.9 lb)Certifications- CEWarranty- 2 years