Learn More
Research Products International Corp LED Digital Orbital Shaker
SDP
Catalog No. 502388430
Encompass
LED Digital Orbital Shaker SLx is ideal for applications including bacterial suspension, solubility studies, growth of bacteria/yeast, extraction procedures, washing, diagnostics tests, hybridization, staining and destaining, mixing and extracting. Suitable for mixing liquids in a variety of vessels including micro-tubes, Petri dishes, cell culture flasks, Erlenmeyer flasks, tubes, microplates and more. Unit features speed and timer LED for independent display of time and speed; continuous or timed operation; anti-slip platform for culture flasks and straps to secure tubes. Brushless motor offers maximum longevity, stable operation and low vibration.
Cold room or incubator compatible
Continuous or timed operation
Two year warranty
Catalog No. 50-238-8430 Supplier Research Products International Corp Supplier No. 400703
LED Digital Orbital Shaker SLx is ideal for applications including bacterial suspension, solubility studies, growth of bacteria/yeast, extraction procedures, washing, diagnostics tests, hybridization, staining and destaining, mixing and extracting. Suitable for mixing liquids in a variety of vessels including micro-tubes, Petri dishes, cell culture flasks, Erlenmeyer flasks, tubes, microplates and more. Unit features speed and timer LED for independent display of time and speed; continuous or timed operation; anti-slip platform for culture flasks and straps to secure tubes. Brushless motor offers maximum longevity, stable operation and low vibration.
Cold room or incubator compatible
Continuous or timed operation
Two year warranty