Learn More
Microscope World SCHOTT KL 1600 LED Dual Pipe Light Illuminator
SDP
Catalog No. NC2965459
Encompass_Preferred
SCHOTT KL 1600 LED Cold Light Source Dual Pipe Light System includes a dual gooseneck light guide, 2 focusing light guide attachments, and a KL 1600 LED cold light source.Gooseneck - Dual Flexible and Obedient 600mm (23.62inch) long, tip is 6mm (0.24inch) diameter.Focusing - Focusing attachments screw onto each dual light guide.Color Temperature - 5600KLuminous Flux - Up to 6802 lm light flux.Voltage - 100~240V, 50-60Hz.Light Box - Intensity control 0-100%, and filter slot with 2-position filter slider.Light Box Dimensions / Weight - 114mm x 231mm x 137mm (4.5inch x 9.1inch x 5.4inch), 2.45kg (5.4 lb)Certifications - CEWarranty - 2 years
Catalog No. NC2965459 Supplier Microscope World Supplier No. KL1600DUAL
SCHOTT KL 1600 LED Cold Light Source Dual Pipe Light System includes a dual gooseneck light guide, 2 focusing light guide attachments, and a KL 1600 LED cold light source.Gooseneck - Dual Flexible and Obedient 600mm (23.62inch) long, tip is 6mm (0.24inch) diameter.Focusing - Focusing attachments screw onto each dual light guide.Color Temperature - 5600KLuminous Flux - Up to 6802 lm light flux.Voltage - 100~240V, 50-60Hz.Light Box - Intensity control 0-100%, and filter slot with 2-position filter slider.Light Box Dimensions / Weight - 114mm x 231mm x 137mm (4.5inch x 9.1inch x 5.4inch), 2.45kg (5.4 lb)Certifications - CEWarranty - 2 years